Gene Transcripts

Gene Symbol
Slc30a4
Entrez Gene ID
22785
Gene Name
solute carrier family 30 (zinc transporter), member 4
Gene Aliases
Znt4, lm, znT-4
Chromosome Location
Chr. 2: 122681233 - 122702663 on Build GRCm38
UniGene ID
Mm.27801
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290993.1 NP_001277922.1 2 721
NM_011774.3 NP_035904.2 3 868
GenBank mRNA AF003747.1 - 3 669
AK166106.1 - 2 723
BC117997.1 - 3 726 40095367
CV556160.1 - 1 32

View Full Product Details