Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011782.2 | NP_035912.2 | 7 | 3011 | |
GenBank mRNA | AF140673.1 | - | 7 | 2170 | |
AK044746.1 | - | 7 | 1233 | ||
AK082475.1 | - | 7 | 1233 | ||
BC138619.1 | - | 7 | 2195 | 8861639 | |
BC138620.1 | - | 7 | 2195 | 8861640 |