Gene Transcripts

Gene Symbol
Psmc4
Entrez Gene ID
23996
Gene Name
proteasome (prosome, macropain) 26S subunit, ATPase, 4
Gene Aliases
CIP21, MIP224
Chromosome Location
Chr. 7: 28041702 - 28050092 on Build GRCm38
UniGene ID
Mm.29582
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011874.2 NP_036004.2 11 1286
GenBank mRNA AK050677.1 - 11 1264
AK077507.1 - 11 1249
AK153991.1 - 11 1269
AK160644.1 - 11 1248
AK160718.1 - 11 1240
AK167041.1 - 11 1231
AK167528.1 - 11 1241
AK169224.1 - 11 1249
AK169244.1 - 11 1748
BC012411.1 - 3 508
BC012708.1 - 11 1247 4211677
BC092265.1 - 11 1230 3671250
BC094063.1 - 6 604
L76223.1 - 11 1237

View Full Product Details