Gene Transcripts

Gene Symbol
Srp54a
Entrez Gene ID
24067
Gene Name
signal recognition particle 54A
Gene Aliases
54kDa, Srp54
Chromosome Location
Chr. 12: 55080496 - 55115367 on Build GRCm38
UniGene ID
Mm.471968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011899.4 NP_036029.2 16 2093
GenBank mRNA AK011928.1 - 16 2057
BC019683.1 - 16 2022 3708250

Gene Symbol
Srp54b
Entrez Gene ID
665155
Gene Name
signal recognition particle 54B
Gene Aliases
Chromosome Location
Chr. 12: 55155104 - 55189213 on Build GRCm38
UniGene ID
Mm.12848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100109.1 NP_001093579.1 16 2329
GenBank mRNA AK136033.1 - 16 2330
X16319.1 - 2058

Gene Symbol
Srp54c
Entrez Gene ID
100101806
Gene Name
signal recognition particle 54C
Gene Aliases
E330029E12Rik
Chromosome Location
Chr. 12: 55230105 - 55279334 on Build GRCm38
UniGene ID
Mm.471968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100110.1 NP_001093580.1 2019
GenBank mRNA AK090018.1 - 2023

View Full Product Details