Gene Transcripts

Gene Symbol
Ctss
Entrez Gene ID
50654
Gene Name
cathepsin S
Gene Aliases
Chromosome Location
Chr. 2: 197655780 - 197679768 on Build Rnor_6.0
UniGene ID
Rn.11347
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017320.1 NP_059016.1 282
GenBank mRNA FQ218804.1 - 357
L03201.1 - 282

View Full Product Details