Gene Transcripts

Gene Symbol
Zic1
Entrez Gene ID
64618
Gene Name
Zic family member 1
Gene Aliases
Chromosome Location
Chr. 8: 98733715 - 98738914 on Build Rnor_6.0
UniGene ID
Rn.161923
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022677.1 NP_073168.1 1 428
XM_008766454.1 XP_008764676.1 2 796
GenBank mRNA AF221839.1 - 1 428

View Full Product Details