Gene Transcripts

Gene Symbol
Mmp9
Entrez Gene ID
81687
Gene Name
matrix metallopeptidase 9
Gene Aliases
Chromosome Location
Chr. 3: 161413410 - 161421473 on Build Rnor_6.0
UniGene ID
Rn.10209
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031055.1 NP_112317.1 1 83
GenBank mRNA U24441.1 - 1 83
U36476.1 - 1 83

View Full Product Details