Gene Transcripts

Gene Symbol
Msh2
Entrez Gene ID
81709
Gene Name
mutS homolog 2
Gene Aliases
Chromosome Location
Chr. 6: 11215951 - 11274916 on Build Rnor_6.0
UniGene ID
Rn.3174
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031058.1 NP_112320.1 1 187
GenBank mRNA BC161846.1 - 1 203
X93591.1 - 1 187

View Full Product Details