Gene Transcripts
- Gene Symbol
- Soat1
- Entrez Gene ID
- 81782
- Gene Name
- sterol O-acyltransferase 1
- Gene Aliases
- Acat-1
- Chromosome Location
- Chr. 13: 74032622 - 74077759 on Build Rnor_6.0
- UniGene ID
- Rn.59
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031118.1 |
NP_112380.1
|
2 |
169 |
|
GenBank mRNA |
D86373.1 |
- |
2 |
169 |
|
View Full Product Details