Gene Transcripts

Gene Symbol
F2rl2
Entrez Gene ID
29636
Gene Name
coagulation factor II (thrombin) receptor-like 2
Gene Aliases
PAR3
Chromosome Location
Chr. 2: 26240385 - 26245533 on Build Rnor_6.0
UniGene ID
Rn.206020
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053313.1 NP_445765.1 2 285
GenBank mRNA AF310076.1 - 2 285

View Full Product Details