Gene Transcripts

Gene Symbol
Clic5
Entrez Gene ID
94272
Gene Name
chloride intracellular channel 5
Gene Aliases
Chromosome Location
Chr. 9: 19121676 - 19372673 on Build Rnor_6.0
UniGene ID
Rn.1838
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053603.2 NP_446055.1 4 637
GenBank mRNA AF323174.1 - 4 645

View Full Product Details