Gene Transcripts

Gene Symbol
Xpnpep2
Entrez Gene ID
117522
Gene Name
X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
Gene Aliases
Chromosome Location
Chr. X: 134940699 - 134969874 on Build Rnor_6.0
UniGene ID
Rn.56142
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057155.3 NP_476496.1 7 648
XM_008773513.1 XP_008771735.1 8 1088
XM_008773514.1 XP_008771736.1 7 951
GenBank mRNA AF359355.1 - 7 648
BC074017.1 - 7 912 7098467

View Full Product Details