Gene Transcripts
- Gene Symbol
- Daxx
- Entrez Gene ID
- 140926
- Gene Name
- death-domain associated protein
- Gene Aliases
- Chromosome Location
- Chr. 20: 5480103 - 5485957 on Build Rnor_6.0
- UniGene ID
- Rn.870
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080891.2 |
NP_543167.2
|
6, 7 |
2014 |
|
XM_006256103.2 |
XP_006256165.1
|
6, 7 |
2796 |
|
XM_006256104.2 |
XP_006256166.1
|
6, 7 |
2470 |
|
XM_006256105.2 |
XP_006256167.1
|
6, 7 |
2366 |
|
XM_006256106.2 |
XP_006256168.1
|
6, 7 |
2032 |
|
XM_006256107.2 |
XP_006256169.1
|
6, 7 |
2136 |
|
XM_006256108.2 |
XP_006256170.1
|
6, 7 |
2016 |
|
XM_006256109.1 |
XP_006256171.1
|
5, 6 |
2889 |
|
XM_006256110.2 |
XP_006256172.1
|
6, 7 |
2015 |
|
XM_008772704.1 |
XP_008770926.1
|
5, 6 |
2889 |
|
GenBank mRNA |
AB064671.1 |
- |
5, 6 |
1921 |
|
BC128729.1 |
- |
6, 7 |
2014 |
8366304 |
FQ223571.1 |
- |
6, 7 |
1985 |
|
View Full Product Details