Gene Transcripts
- Gene Symbol
- Mgea5
- Entrez Gene ID
- 154968
- Gene Name
- meningioma expressed antigen 5 (hyaluronidase)
- Gene Aliases
- Ncoat, O-GLCNACASE, OGA
- Chromosome Location
- Chr. 1: 265508619 - 265541300 on Build Rnor_6.0
- UniGene ID
- Rn.162539
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_131904.1 |
NP_571979.1
|
3, 4 |
359 |
|
GenBank mRNA |
AY039679.1 |
- |
3, 4 |
359 |
|
View Full Product Details