Gene Transcripts

Gene Symbol
Pou3f3
Entrez Gene ID
192109
Gene Name
POU class 3 homeobox 3
Gene Aliases
Brn1, RHS1
Chromosome Location
Chr. 9: 49479014 - 49482246 on Build Rnor_6.0
UniGene ID
Rn.11354
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138837.1 NP_620192.1 1 1843
GenBank mRNA AJ001641.1 - 1 1843

View Full Product Details