Gene Transcripts

Gene Symbol
Cct4
Entrez Gene ID
29374
Gene Name
chaperonin containing Tcp1, subunit 4 (delta)
Gene Aliases
Chromosome Location
Chr. 14: 107767392 - 107780270 on Build Rnor_6.0
UniGene ID
Rn.97889
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182814.2 NP_877966.1 5 645
GenBank mRNA AY223861.1 - 5 511
BC079283.1 - 5 616 7134210

View Full Product Details