Gene Transcripts

Gene Symbol
Art2b
Entrez Gene ID
293152
Gene Name
ADP-ribosyltransferase 2b
Gene Aliases
ARTC2, Ag-F, Art2, Art2a, LY2, Ly-2, PtaA, RT6.1, RT6.2, Rt6
Chromosome Location
Chr. 1: 166667304 - 166685081 on Build Rnor_6.0
UniGene ID
Rn.107075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198735.2 NP_942030.1 2 245
XM_008759694.1 XP_008757916.1 2 473
XM_008759695.1 XP_008757917.1 2 473
XM_008759696.1 XP_008757918.1 2 1094
XM_008759697.1 XP_008757919.1 3 1462
XM_008759698.1 XP_008757920.1 5 834
XM_008759699.1 XP_008757921.1 2 320
XM_008759700.1 XP_008757922.1 4 565
XM_008759701.1 XP_008757923.1 2 388
XM_008759702.1 XP_008757924.1 2 1094
GenBank mRNA BC099070.1 - 2 245
FQ227811.1 - 1 18
FQ230306.1 - 4 581
M85193.1 - 2 222
X52082.1 - 2 215
X99122.1 - 5 839
X99123.1 - 3 295
X99124.1 - 3 312

View Full Product Details