Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318029.1 | NP_001304958.1 | 4 | 1842 | |
NM_176875.3 | NP_795344.1 | 5 | 2094 | ||
XM_005253210.1 | XP_005253267.1 | 4 | 2364 | ||
XM_017018516.1 | XP_016874005.1 | 5 | 2580 | ||
GenBank mRNA | AF239668.1 | - | 4 | 2049 | |
AK226020.1 | - | 6 | 1856 | ||
BC000740.2 | - | 5 | 1895 | 3504160 | |
CD013884.1 | - | 4 | 1710 | ||
D13305.1 | - | 5 | 2031 | ||
L04473.1 | - | 5 | 1838 | ||
S70057.1 | - | 5 | 2020 |