Gene Transcripts
- Gene Symbol
- Pggt1b
- Entrez Gene ID
- 81746
- Gene Name
- protein geranylgeranyltransferase type I, beta subunit
- Gene Aliases
- Chromosome Location
- Chr. 18: 40173236 - 40218225 on Build Rnor_6.0
- UniGene ID
- Rn.10031
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031082.1 |
NP_112344.1
|
1, 2 |
154 |
|
GenBank mRNA |
L24116.1 |
- |
1, 2 |
154 |
|
View Full Product Details