Gene Transcripts

Gene Symbol
Spag4
Entrez Gene ID
83623
Gene Name
sperm associated antigen 4
Gene Aliases
Chromosome Location
Chr. 3: 151609602 - 151613946 on Build Rnor_6.0
UniGene ID
Rn.28620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031792.1 NP_113980.1 3 549
XM_008762386.1 XP_008760608.1 3 283
GenBank mRNA AF043345.2 - 3 549

Gene Symbol
LOC100911109
Entrez Gene ID
100911109
Gene Name
sperm-associated antigen 4 protein-like
Gene Aliases
Chromosome Location
Chr. 3: 152143565 - 152148051 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749611.3 XP_003749659.1 691
XM_008762411.1 XP_008760633.1 283

View Full Product Details