Gene Transcripts

Gene Symbol
Higd1a
Entrez Gene ID
140937
Gene Name
HIG1 hypoxia inducible domain family, member 1A
Gene Aliases
Hig1
Chromosome Location
Chr. 8: 130482492 - 130491888 on Build Rnor_6.0
UniGene ID
Rn.2084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080902.4 NP_543178.2 4 356
GenBank mRNA AY062253.3 - 4 415
BC059118.1 - 4 360 6890532

View Full Product Details