Gene Transcripts

Gene Symbol
Slc40a1
Entrez Gene ID
170840
Gene Name
solute carrier family 40 (iron-regulated transporter), member 1
Gene Aliases
Fpn1, Slc11a3, Slc39a1
Chromosome Location
Chr. 9: 52894365 - 52912293 on Build Rnor_6.0
UniGene ID
Rn.15324
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133315.2 NP_579849.2 2 369
GenBank mRNA AF394785.3 - 2 314
CB808602.1 - 2 380
FQ226522.1 - 1 29
U76714.1 - 2 384

View Full Product Details