Gene Transcripts

Gene Symbol
Cx3cr1
Entrez Gene ID
171056
Gene Name
chemokine (C-X3-C motif) receptor 1
Gene Aliases
Rbs11
Chromosome Location
Chr. 8: 128738689 - 128755376 on Build Rnor_6.0
UniGene ID
Rn.10482
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133534.1 NP_598218.1 2 387
XM_008766668.1 XP_008764890.1 2 702
GenBank mRNA U04808.1 - 2 387

View Full Product Details