Gene Transcripts

Gene Symbol
KIF22
Entrez Gene ID
3835
Gene Name
kinesin family member 22
Gene Aliases
A-328A3.2, KID, KNSL4, OBP, OBP-1, OBP-2, SEMDJL2
Chromosome Location
Chr. 16: 29790062 - 29805385 on Build GRCh38
UniGene ID
Hs.612151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256269.1 NP_001243198.1 3 353
NM_001256270.1 NP_001243199.1 2 441
NM_007317.2 NP_015556.1 2 304
GenBank mRNA AB017430.2 - 2 281
AK223431.1 - 2 290
AK294380.1 - 2 680
AK297893.1 - 2 284
AK312234.1 - 2 295
AK316050.1 - 2 441
AK316389.1 - 3 312
BC004352.1 - 2 276 3535435
BC028155.1 - 2 262 5241557
BT007259.1 - 2 257
DC370294.1 - 2 304

View Full Product Details