Gene Transcripts

Gene Symbol
Mcfd2
Entrez Gene ID
246117
Gene Name
multiple coagulation factor deficiency 2
Gene Aliases
Sdnsf
Chromosome Location
Chr. 6: 10887303 - 10899221 on Build Rnor_6.0
UniGene ID
Rn.7871
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139253.1 NP_640346.1 4 1336
GenBank mRNA AF475282.1 - 4 1336
BC071179.1 - 4 1330 7100216

View Full Product Details