Gene Transcripts
- Gene Symbol
- Drgx
- Entrez Gene ID
- 252880
- Gene Name
- dorsal root ganglia homeobox
- Gene Aliases
- Drg11, Prrxl1
- Chromosome Location
- Chr. 16: 8823872 - 8853430 on Build Rnor_6.0
- UniGene ID
- Rn.10189
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145767.1 |
NP_665710.1
|
7 |
1404 |
|
GenBank mRNA |
U29174.1 |
- |
7 |
1404 |
|
View Full Product Details