Gene Transcripts
- Gene Symbol
- Vom1r90
- Entrez Gene ID
- 266771
- Gene Name
- vomeronasal 1 receptor 90
- Gene Aliases
- V1ra16
- Chromosome Location
- Chr. 4: 121675261 - 121684264 on Build Rnor_6.0
- UniGene ID
- Rn.234590
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153729.2 |
NP_714951.1
|
3 |
1288 |
|
GenBank mRNA |
U36898.1 |
- |
3 |
1308 |
|
View Full Product Details