Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022236.1 | NP_071572.1 | 2 | 402 | |
GenBank mRNA | AB027155.1 | - | 2 | 402 | |
AB027156.1 | - | 737 | |||
AY462091.1 | - | 368 | |||
AY462092.1 | - | 476 | |||
AY462093.1 | - | 514 | |||
AY462095.1 | - | 649 |