Gene Transcripts

Gene Symbol
Gabrq
Entrez Gene ID
65187
Gene Name
gamma-aminobutyric acid (GABA) A receptor, theta
Gene Aliases
Chromosome Location
Chr. X: 152852243 - 152865049 on Build Rnor_6.0
UniGene ID
Rn.81067
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031733.1 NP_113921.1 9 1578
XM_006229516.2 XP_006229578.1 10 1742
GenBank mRNA AF189261.1 - 1 192
AF419333.1 - 9 1578

View Full Product Details