Gene Transcripts

Gene Symbol
TMC6
Entrez Gene ID
11322
Gene Name
transmembrane channel like 6
Gene Aliases
EV1, EVER1, EVIN1, LAK-4P
Chromosome Location
Chr. 17: 78108775 - 78132427 on Build GRCh38
UniGene ID
Hs.632227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127198.2 NP_001120670.1 19 2551
NM_001321185.1 NP_001308114.1 19 2504
NM_007267.7 NP_009198.4 19 2455
XM_005256995.1 XP_005257052.1 19 2580
XM_011524255.1 XP_011522557.1 18 2482
XM_011524256.1 XP_011522558.1 18 2356
XM_011524257.2 XP_011522559.1 13 1600
XM_011524258.1 XP_011522560.1 10 1432
XM_017024107.1 XP_016879596.1 18 2349
XM_017024108.1 XP_016879597.1 18 2302
XM_017024109.1 XP_016879598.1 17 2271
GenBank mRNA AB002405.2 - 11 1263
AK074065.1 - 19 2454
AK310734.1 - 18 2875
AY057379.1 - 18 2294
AY099356.1 - 20 2453
AY099357.1 - 12 1400
AY236496.1 - 18 2420
BC018346.1 - 12 1419
BC023597.2 - 19 2435 4566166
BC035648.1 - 12 1475 5575949
DQ104440.1 - 9 1031

View Full Product Details