Gene Transcripts
- Gene Symbol
- Dll1
- Entrez Gene ID
- 84010
- Gene Name
- delta-like 1 (Drosophila)
- Gene Aliases
- Chromosome Location
- Chr. 1: 57318621 - 57326732 on Build Rnor_6.0
- UniGene ID
- Rn.10628
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032063.2 |
NP_114452.1
|
6, 7 |
1155 |
|
GenBank mRNA |
U78889.2 |
- |
6, 7 |
1155 |
|
View Full Product Details