Gene Transcripts

Gene Symbol
Retnla
Entrez Gene ID
81712
Gene Name
resistin like alpha
Gene Aliases
Himf
Chromosome Location
Chr. 11: 54543729 - 54545480 on Build Rnor_6.0
UniGene ID
Rn.12120
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053333.1 NP_445785.1 3 396
XM_006248283.2 XP_006248345.1 4 488
GenBank mRNA AF323085.1 - 3 396

View Full Product Details