Gene Transcripts

Gene Symbol
STAP1
Entrez Gene ID
26228
Gene Name
signal transducing adaptor family member 1
Gene Aliases
BRDG1, STAP-1
Chromosome Location
Chr. 4: 67558697 - 67607341 on Build GRCh38
UniGene ID
Hs.435579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317769.1 NP_001304698.1 2 272
NM_012108.3 NP_036240.1 2 272
XM_017008018.1 XP_016863507.1 2 272
GenBank mRNA AB023483.1 - 2 242
AK313676.1 - 2 221
BC014958.1 - 2 191 4851485

View Full Product Details