Gene Transcripts

Gene Symbol
Acsl3
Entrez Gene ID
114024
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
Acs3, Facl3
Chromosome Location
Chr. 9: 84569601 - 84593565 on Build Rnor_6.0
UniGene ID
Rn.54820
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057107.1 NP_476448.1 3 679
GenBank mRNA D30666.1 - 3 679
FQ221868.1 - 889

View Full Product Details