Gene Transcripts
- Gene Symbol
- P2rx5
- Entrez Gene ID
- 113995
- Gene Name
- purinergic receptor P2X, ligand-gated ion channel, 5
- Gene Aliases
- P2x5
- Chromosome Location
- Chr. 10: 59725435 - 59737126 on Build Rnor_6.0
- UniGene ID
- Rn.10257
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080780.2 |
NP_542958.2
|
6 |
758 |
|
GenBank mRNA |
X92069.1 |
- |
6 |
758 |
|
View Full Product Details