Gene Transcripts

Gene Symbol
Dgka
Entrez Gene ID
140866
Gene Name
diacylglycerol kinase, alpha
Gene Aliases
Dagk1
Chromosome Location
Chr. 7: 3143826 - 3170768 on Build Rnor_6.0
UniGene ID
Rn.1840
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080787.1 NP_542965.1 4, 5 786
XM_008765012.1 XP_008763234.1 4, 5 1566
XM_008765013.1 XP_008763235.1 4, 5 1566
XM_008765014.1 XP_008763236.1 4, 5 1566
XM_008765015.1 XP_008763237.1 4, 5 1408
XM_008765016.1 XP_008763238.1 4, 5 649
XM_008765017.1 XP_008763239.1 4, 5 1566
XM_008765018.1 XP_008763240.1 4, 5 1566
XM_008765019.1 XP_008763241.1 4, 5 1565
XM_008765020.1 XP_008763242.1 4, 5 1402
GenBank mRNA FQ209812.1 - 4, 5 590
S49760.1 - 4, 5 786

View Full Product Details