Gene Transcripts

Gene Symbol
Ikbkap
Entrez Gene ID
140934
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
Chromosome Location
Chr. 5: 73503406 - 73552798 on Build Rnor_6.0
UniGene ID
Rn.162816
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080899.1 NP_543175.1 4 438
GenBank mRNA AF388201.1 - 4 438

View Full Product Details