Gene Transcripts
- Gene Symbol
- Ikbkap
- Entrez Gene ID
- 140934
- Gene Name
- inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
- Gene Aliases
- Chromosome Location
- Chr. 5: 73503406 - 73552798 on Build Rnor_6.0
- UniGene ID
- Rn.162816
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080899.1 |
NP_543175.1
|
4, 5 |
476 |
|
GenBank mRNA |
AF388201.1 |
- |
4, 5 |
476 |
|
View Full Product Details