Gene Transcripts

Gene Symbol
Gpnmb
Entrez Gene ID
113955
Gene Name
glycoprotein (transmembrane) nmb
Gene Aliases
Chromosome Location
Chr. 4: 78694447 - 78715685 on Build Rnor_6.0
UniGene ID
Rn.13778
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133298.1 NP_579832.1 5, 6 806
GenBank mRNA AF184983.2 - 5, 6 806
BC061725.1 - 5, 6 749 5598392

View Full Product Details