Gene Transcripts
- Gene Symbol
- Mutyh
- Entrez Gene ID
- 170841
- Gene Name
- mutY homolog
- Gene Aliases
- Myh
- Chromosome Location
- Chr. 5: 135663332 - 135675348 on Build Rnor_6.0
- UniGene ID
- Rn.44045
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133316.1 |
NP_579850.1
|
14 |
1324 |
|
GenBank mRNA |
AF478683.1 |
- |
14 |
1324 |
|
View Full Product Details