Gene Transcripts

Gene Symbol
Folr1
Entrez Gene ID
171049
Gene Name
folate receptor 1 (adult)
Gene Aliases
Fbp1
Chromosome Location
Chr. 1: 166934457 - 166943592 on Build Rnor_6.0
UniGene ID
Rn.211829
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133527.1 NP_598211.1 3 260
XM_006229842.2 XP_006229904.1 3 381
GenBank mRNA AF219904.1 - 3 260
BC157811.1 - 2 243

View Full Product Details