Gene Transcripts
- Gene Symbol
- FBXO3
- Entrez Gene ID
- 26273
- Gene Name
- F-box protein 3
- Gene Aliases
- FBA, FBX3
- Chromosome Location
- Chr. 11: 33740937 - 33774531 on Build GRCh38
- UniGene ID
- Hs.406787
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012175.3 |
NP_036307.2
|
2, 3 |
231 |
|
NM_033406.2 |
NP_208385.1
|
2, 3 |
231 |
|
XM_011519980.1 |
XP_011518282.1
|
2, 3 |
237 |
|
XM_011519981.2 |
XP_011518283.1
|
2, 3 |
237 |
|
XM_011519982.1 |
XP_011518284.1
|
2, 3 |
210 |
|
XM_017017533.1 |
XP_016873022.1
|
2, 3 |
210 |
|
GenBank mRNA |
AB209039.1 |
- |
1, 2 |
3616 |
|
AF174595.1 |
- |
2, 3 |
191 |
|
AF176702.1 |
- |
2, 3 |
188 |
|
AK001943.1 |
- |
2, 3 |
222 |
|
AK128454.1 |
- |
2, 3 |
209 |
|
AK304866.1 |
- |
2, 3 |
226 |
|
AL136625.1 |
- |
2, 3 |
214 |
|
AL832826.1 |
- |
1, 2 |
903 |
|
BC039291.1 |
- |
2, 3 |
231 |
|
BC046110.1 |
- |
2, 3 |
217 |
5549548 |
BG716664.1 |
- |
2, 3 |
234 |
|
View Full Product Details