Gene Transcripts

Gene Symbol
Tnfsf15
Entrez Gene ID
252878
Gene Name
tumor necrosis factor (ligand) superfamily, member 15
Gene Aliases
Chromosome Location
Chr. 5: 79553935 - 79569974 on Build Rnor_6.0
UniGene ID
Rn.84873
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145765.1 NP_665708.1 3, 4 305
GenBank mRNA AF520787.1 - 3, 4 305

View Full Product Details