Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_171996.3 | NP_741993.1 | 5 | 444 | |
XM_006246024.2 | XP_006246086.1 | 6 | 644 | ||
GenBank mRNA | AF044574.1 | - | 5 | 366 | |
BC070959.1 | - | 5 | 445 | 7097883 | |
FQ218511.1 | - | 5 | 419 |