Gene Transcripts

Gene Symbol
Angptl4
Entrez Gene ID
362850
Gene Name
angiopoietin-like 4
Gene Aliases
Chromosome Location
Chr. 7: 18627814 - 18634043 on Build Rnor_6.0
UniGene ID
Rn.119611
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199115.2 NP_954546.1 2 520
GenBank mRNA AY393999.1 - 2 380
BC078944.1 - 2 520 7109431

View Full Product Details