Gene Transcripts

Gene Symbol
Htr4
Entrez Gene ID
25324
Gene Name
5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled
Gene Aliases
Chromosome Location
Chr. 18: 57637297 - 57820258 on Build Rnor_6.0
UniGene ID
Rn.10094
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012853.1 NP_036985.1 5 797
XM_006254793.2 XP_006254855.1 6 1336
XM_008772089.1 XP_008770311.1 4 2462
XM_008772090.1 XP_008770312.1 4 2462
GenBank mRNA AJ011370.1 - 6 877
AM712909.1 - 5 777
AM712910.1 - 5 777
AM712911.1 - 5 777
U20906.1 - 5 797
U20907.1 - 5 797

View Full Product Details