Gene Transcripts

Gene Symbol
Faslg
Entrez Gene ID
25385
Gene Name
Fas ligand (TNF superfamily, member 6)
Gene Aliases
Apt1Lg1, CD95-L, Fasl, Tnfsf6
Chromosome Location
Chr. 13: 79696813 - 79716821 on Build Rnor_6.0
UniGene ID
Rn.9725
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012908.1 NP_037040.1 1 399
XM_006250148.2 XP_006250210.1 4 1038
XM_008769607.1 XP_008767829.1 5 1273
GenBank mRNA U03470.1 - 1 399

View Full Product Details