Gene Transcripts
- Gene Symbol
- AUP1
- Entrez Gene ID
- 550
- Gene Name
- ancient ubiquitous protein 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 74526648 - 74529897 on Build GRCh38
- UniGene ID
- Hs.411480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181575.4 |
NP_853553.1
|
11, 12 |
1472 |
|
NR_126510.1 |
|
11, 12 |
1534 |
|
NR_126511.1 |
|
10, 11 |
1668 |
|
XM_005264392.3 |
XP_005264449.1
|
10, 11 |
1417 |
|
GenBank mRNA |
AF100746.1 |
- |
11, 12 |
1245 |
|
AF100753.1 |
- |
11, 12 |
1272 |
|
AF100754.1 |
- |
10, 11 |
1470 |
|
AF165515.1 |
- |
11, 12 |
1252 |
|
AK023983.1 |
- |
11, 12 |
1312 |
|
AK055566.1 |
- |
7, 8 |
2104 |
|
AK293121.1 |
- |
11, 12 |
1291 |
|
AK304886.1 |
- |
10, 11 |
1414 |
|
BC001658.1 |
- |
11, 12 |
1397 |
3050204 |
BC033646.2 |
- |
11, 12 |
1287 |
|
CR749661.1 |
- |
10, 11 |
1405 |
|
View Full Product Details