Gene Transcripts
- Gene Symbol
- FBXO3
- Entrez Gene ID
- 26273
- Gene Name
- F-box protein 3
- Gene Aliases
- FBA, FBX3
- Chromosome Location
- Chr. 11: 33740937 - 33774531 on Build GRCh38
- UniGene ID
- Hs.406787
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012175.3 |
NP_036307.2
|
8, 9 |
968 |
|
NM_033406.2 |
NP_208385.1
|
8, 9 |
968 |
|
XM_011519980.1 |
XP_011518282.1
|
8, 9 |
974 |
|
XM_011519981.2 |
XP_011518283.1
|
8, 9 |
974 |
|
XM_011519982.1 |
XP_011518284.1
|
9, 10 |
1028 |
|
XM_017017533.1 |
XP_016873022.1
|
9, 10 |
1028 |
|
GenBank mRNA |
AB209039.1 |
- |
7, 8 |
4353 |
|
AF176702.1 |
- |
8, 9 |
925 |
|
AK001943.1 |
- |
8, 9 |
959 |
|
AK128454.1 |
- |
8, 9 |
946 |
|
AK300721.1 |
- |
3, 4 |
506 |
|
AK304866.1 |
- |
7, 8 |
758 |
|
AK316286.1 |
- |
3, 4 |
274 |
|
AL136625.1 |
- |
8, 9 |
950 |
|
AL832826.1 |
- |
7, 8 |
1640 |
|
BC039291.1 |
- |
8, 9 |
2313 |
|
BC046110.1 |
- |
8, 9 |
954 |
5549548 |
View Full Product Details