Gene Transcripts

Gene Symbol
Adora2b
Entrez Gene ID
29316
Gene Name
adenosine A2B receptor
Gene Aliases
Chromosome Location
Chr. 10: 48569563 - 48586366 on Build Rnor_6.0
UniGene ID
Rn.89544
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017161.1 NP_058857.1 1 215
GenBank mRNA BC161854.1 - 1 223
M91466.1 - 1 215

View Full Product Details